N-Ethylmaleimide (NEM) 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00355-09
L00355-18
CAAAAL00355-09EA
169.06
CAD
CAAAAL00355-09
CAAAAL00355-18
N-Ethylmaleimide (NEM) 98+%
N-Ethylmaleimide
Cysteine alkylating agent
Purity: 98+%
CAS No.: 128-53-0
Molecular Formula: C6H7NO2
Formula Weight: 125.13
Flash Point: 73°(163°F)
Odor: Pungent
Boiling Point: 209-211°
Beilstein Reference: 112448
Merck Reference: 14,3822
UN No.: 2928
MDL No.: MFCD00005509
Formula:
C₆H₇NO₂ Boiling Pt: 209…211 °C Melting Pt: 43…46 °C Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005509 CAS Number: 128-53-0 EINECS: 204-892-4 UN: 2928 ADR: 6.1,II Merck Index: 14,3822 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...