Triethyl phosphite 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00339-AE
L00339-AP
CAAAAL00339-AEEA
47.24
CAD
CAAAAL00339-AE
CAAAAL00339-AP
Triethyl phosphite 98%
Triethyl phosphite
Purity: 98%
CAS No.: 122-52-1
Molecular Formula: C6H15O3P
Formula Weight: 166.16
Flash Point: 43°(109°F)
Refractive Index: 1.4130
Odor: Unpleasant
Melting Point: -112°
Boiling Point: 155-157°
Beilstein Reference: 956578
UN No.: 2323
MDL No.: MFCD00009084
Formula:
C₆H₁₅O₃P Boiling Pt: 155…157 °C Melting Pt: –112 °C Density: 0.958 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009084 CAS Number: 122-52-1 EINECS: 204-552-5 UN: 2323 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...