4-Nitrophenyl acetate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
P-Acetoxynitrobenzene, 1-Acetoxy-4-nitrobenzene
L00314-09
L00314-18
CAAAAL00314-09EA
61.78
CAD
CAAAAL00314-09
CAAAAL00314-18
4-Nitrophenyl acetate 98+%
4-Nitrophenyl acetate
Purity: 98+%
CAS No.: 830-03-5
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Melting Point: 77-79°
Beilstein Reference: 515874
Formula:
C₈H₇NO₄ MW: 181.15 g/mol Melting Pt: 77…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007326 CAS Number: 830-03-5 EINECS: 212-593-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...