tert-Butyl carbamate 98+%
Supplier: Thermo Scientific Chemicals
L00259-06
L00259-14
CAAAAL00259-03EA
39.9
CAD
CAAAAL00259-03
CAAAAL00259-06
CAAAAL00259-14
tert-Butyl carbamate 98+%
tert-Butyl carbamate
Purity: 98+%
CAS No.: 4248-19-5
Molecular Formula: H2NCO2C(CH3)3
Formula Weight: 117.15
Melting Point: 106-110°
Beilstein Reference: 1744500
Formula:
C₅H₁₁NO₂ Melting Pt: 106…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007962 CAS Number: 4248-19-5 EINECS: 224-209-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...