3,4-Epoxy-1-butene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00238-03
L00238-06
L00238-14
CAAAAL00238-03EA
97.49
CAD
CAAAAL00238-03
CAAAAL00238-06
CAAAAL00238-14
3,4-Epoxy-1-butene 98%
3,4-Epoxy-1-butene
Purity: 98%
CAS No.: 930-22-3
Molecular Formula: C4H6O
Formula Weight: 70.09
Storage Temperature: 2°C to 8°C
Flash Point: -50°(-58°F)
Refractive Index: 1.4180
Odor: Recognizable
Melting Point: -135°
Boiling Point: 65-66°
Beilstein Reference: 103170
UN No.: 1993
MDL No.: MFCD00005149
Formula:
C₄H₆O Boiling Pt: 65…66 °C Melting Pt: –135 °C Density: 0.870 Flash Pt: –50 °C (–58 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005149 CAS Number: 930-22-3 EINECS: 213-210-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...