Trichloroacetyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

L00226-04 L00226-09 L00226-18
CAAAAL00226-04EA 63.67 CAD
CAAAAL00226-04 CAAAAL00226-09 CAAAAL00226-18
Trichloroacetyl isocyanate 97%
Trichloroacetyl isocyanate

Purity: 97% CAS No.: 3019-71-4 Molecular Formula: C3Cl3NO2 Formula Weight: 188.40 Storage Temperature: 2°C to 8°C Flash Point: 65°(149°F) Refractive Index: 1.4800 Odor: Acrid Boiling Point: 58-60°/20mm Beilstein Reference: 971201 UN No.: 2927 MDL No.: MFCD00002033

Formula: C₃Cl₃NO₂
MW: 188.4 g/mol
Boiling Pt: 58…60 °C (20 mmHg)
Density: 1.585
Flash Pt: 65 °C (149 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002033
CAS Number: 3019-71-4
EINECS: 221-165-7
UN: 2927
ADR: 6.1 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR