Tetraphenylethylene 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1,1,2,2-Tetraphenylethylene
L00222-06
L00222-14
CAAAAL00222-06EA
99.38
CAD
CAAAAL00222-06
CAAAAL00222-14
Tetraphenylethylene 98%
Tetraphenylethylene
Purity: 98%
CAS No.: 632-51-9
Molecular Formula: C26H20
Formula Weight: 332.45
Melting Point: 222-226°
Boiling Point: 420°
MDL No.: MFCD00004764
Formula:
C₂₆H₂₀ Boiling Pt: 420 °C Melting Pt: 222…226 °C Density: 1.148 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004764 CAS Number: 632-51-9 EINECS: 211-179-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...