N,O-Bis(trimethylsilyl)acetamide 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

L00183-14 L00183-22
CAAAAL00183-14EA 78.86 CAD
CAAAAL00183-14 CAAAAL00183-22
N,O-Bis(trimethylsilyl)acetamide 95%
N,O-Bis(trimethylsilyl)acetamide

Purity: 95% CAS No.: 10416-59-8 Molecular Formula: C8H21NOSi2 Formula Weight: 203.43 Storage Temperature: 2°C to 8°C Flash Point: 42°(107°F) Refractive Index: 1.4170 Melting Point: -24° Boiling Point: 71-73°/35mm Beilstein Reference: 1306669 UN No.: 2920 MDL No.: MFCD00008270

Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
Boiling Pt: 71…73 °C/35 mm
Melting Pt: –24 °C
Density: 0.829
Flash Pt: 42 °C (107 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008270
CAS Number: 10416-59-8
EINECS: 233-892-7
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR