Toluene-p-sulfonic anhydride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Toluenesulfonic anhydride, 4-Methylbenzenesulphonic anhydride, p-Toluenesulphonic anhydride, 4-Methylbenzenesulfonic anhydride

L00160-06 L00160-14
CAAAAL00160-06EA 79.16 CAD
CAAAAL00160-06 CAAAAL00160-14
Toluene-p-sulfonic anhydride 97%
Toluene-p-sulphonic anhydride

Purity: 97% CAS No.: 4124-41-8 Molecular Formula: C14H14O5S2 Formula Weight: 326.38 Melting Point: 125-130° Beilstein Reference: 2223702 UN No.: 3261 MDL No.: MFCD00008548

Formula: C₁₄H₁₄O₅S₂
MW: 326.39 g/mol
Melting Pt: 125…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00008548
CAS Number: 4124-41-8
EINECS: 223-926-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR