tert-Butyl cyanoacetate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl 2-cyanoacetate

L00123-06 L00123-14 L00123-22
CAAAAL00123-06EA 58.62 CAD
CAAAAL00123-06 CAAAAL00123-14 CAAAAL00123-22
tert-Butyl cyanoacetate 98%
tert-Butyl cyanoacetate

Purity: 98% CAS No.: 1116-98-9 Molecular Formula: NCCH2CO2C(CH3)3 Formula Weight: 141.17 Flash Point: 91°(195°F) Refractive Index: 1.4200 Odor: Odorless Boiling Point: 67-68°/1.5mm Beilstein Reference: 1755933 MDL No.: MFCD00001938

Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
Boiling Pt: 67…68 °C/1.5 mm
Density: 0,987
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001938
CAS Number: 1116-98-9
EINECS: 214-243-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR