3-tert-Butylaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(tert-Butyl)aniline, m-tert-Butylaniline
H50698-03
H50698-06
CAAAAH50698-03EA
241.74
CAD
CAAAAH50698-03
CAAAAH50698-06
3-tert-Butylaniline 97%
3-tert-Butylaniline
Purity: 97%
CAS No.: 5369-19-7
Molecular Formula: C10H15N
Formula Weight: 149.23
MDL No.: MFCD00125078
Formula:
C₁₀H₁₅N MW: 149.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00125078 CAS Number: 5369-19-7 EINECS: 226-361-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...