Dimethyl-5-bromoisophthalate 98%

Supplier: Thermo Scientific Chemicals
Warning

H50629-03 H50629-06
CAAAAH50629-03EA 59.57 CAD
CAAAAH50629-03 CAAAAH50629-06
Dimethyl-5-bromoisophthalate 98%
Dimethyl 5-bromoisophthalate

Purity: 98% CAS No.: 51760-21-5 Molecular Formula: C10H9BrO4 Formula Weight: 273.08 Melting Point: 88-90° MDL No.: MFCD00078709

Formula: C₁₀H₉BrO₄
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00078709
CAS Number: 51760-21-5
EINECS: 257-386-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR