5-Bromooxindole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50573-03
H50573-06
CAAAAH50573-03EA
162.74
CAD
CAAAAH50573-03
CAAAAH50573-06
5-Bromooxindole 98%
5-Bromooxindole
Purity: 98%
CAS No.: 20870-78-4
Molecular Formula: C8H6BrNO
Formula Weight: 212.05
MDL No.: MFCD00456998
Formula:
C₈H₆BrNO Storage Temperature: Ambient |
MDL Number:
MFCD00456998 CAS Number: 20870-78-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...