5-Bromo-2-fluoro-m-xylene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50564-03
H50564-06
CAAAAH50564-03EA
73.94
CAD
CAAAAH50564-03
CAAAAH50564-06
5-Bromo-2-fluoro-m-xylene 97%
5-Bromo-2-fluoro-m-xylene
Purity: 97%
CAS No.: 99725-44-7
Molecular Formula: C8H8BrF
Formula Weight: 203.06
Boiling Point: 95°/22mm
MDL No.: MFCD01320701
Formula:
C₈H₈BrF Boiling Pt: 95 °C (22 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD01320701 CAS Number: 99725-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...