3-Chloro-4-hydroxybenzonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

H50434-03 H50434-06
CAAAAH50434-03EA 53.09 CAD
CAAAAH50434-03 CAAAAH50434-06
3-Chloro-4-hydroxybenzonitrile 95%
3-Chloro-4-hydroxybenzonitrile

Purity: 95% CAS No.: 2315-81-3 Molecular Formula: C7H4ClNO Formula Weight: 153.57 Melting Point: 147-149° MDL No.: MFCD01567246

Formula: C₇H₄ClNO
Melting Pt: 147…149 °C
Storage Temperature: Ambient
MDL Number: MFCD01567246
CAS Number: 2315-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR