2,4,5-Trimethylphenol 99%

Supplier: Thermo Scientific Chemicals
Danger

H50427-03 H50427-MD
CAAAAH50427-03EA 451.88 CAD
CAAAAH50427-03 CAAAAH50427-MD
2,4,5-Trimethylphenol 99%
2,4,5-Trimethylphenol

Purity: 99% CAS No.: 496-78-6 Molecular Formula: C9H12O Formula Weight: 136.20 Odor: Phenol-like Melting Point: 72° Boiling Point: 232° UN No.: 2430 MDL No.: MFCD00020050

Formula: C₉H₁₂O
Boiling Pt: 232 °C
Melting Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00020050
CAS Number: 496-78-6
EINECS: 207-832-5
UN: 2430
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR