1,2-Benzenedisulfonyl chloride

Supplier: Thermo Scientific Chemicals
Danger

H50375-03 H50375-06
CAAAAH50375-03EA 369.72 CAD
CAAAAH50375-03 CAAAAH50375-06
1,2-Benzenedisulfonyl chloride
1,2-Benzenedisulphonyl chloride

CAS No.: 6461-76-3 Molecular Formula: C6H4Cl2O4S2 Formula Weight: 275.13 Melting Point: 140-146° UN No.: 3261 MDL No.: MFCD00059836

Formula: C₆H₄Cl₂O₄S₂
Melting Pt: 140…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00059836
CAS Number: 6461-76-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR