3-Methoxy-p-toluidine 99+%

Supplier: Thermo Scientific Chemicals
Warning

H50362-03 H50362-MD
CAAAAH50362-03EA 334.96 CAD
CAAAAH50362-03 CAAAAH50362-MD
3-Methoxy-p-toluidine 99+%
3-Methoxy-p-toluidine

Purity: 99+% CAS No.: 16452-01-0 Molecular Formula: C8H11NO Formula Weight: 137.18 Melting Point: 56-60° UN No.: 2811 MDL No.: MFCD00025371

Formula: C₈H₁₁NO
Boiling Pt: 250 °C (1013 hPa)
Melting Pt: 56…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00025371
CAS Number: 16452-01-0
EINECS: 240-500-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR