2,4,6-Trimethoxybenzyl alcohol 97%
Supplier: Thermo Scientific Chemicals
H50348-03
H50348-MD
CAAAAH50348-03EA
297.04
CAD
CAAAAH50348-03
CAAAAH50348-MD
2,4,6-Trimethoxybenzyl alcohol 97%
2,4,6-Trimethoxybenzyl alcohol
Purity: 97%
CAS No.: 61040-78-6
Molecular Formula: C10H14O4
Formula Weight: 198.22
Melting Point: 60°
MDL No.: MFCD00060356
Formula:
C₁₀H₁₄O₄ MW: 198.22 Melting Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060356 CAS Number: 61040-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...