3-(Difluoromethoxy)benzyl alcohol
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(3-(Difluoromethoxy)phenyl)methanol
H50345-03
H50345-06
CAAAAH50345-03EA
262.28
CAD
CAAAAH50345-03
CAAAAH50345-06
3-(Difluoromethoxy)benzyl alcohol
3-(Difluoromethoxy)benzyl alcohol
CAS No.: 125903-81-3
Molecular Formula: C8H8F2O2
Formula Weight: 174.15
MDL No.: MFCD00236227
Formula:
C₈H₈F₂O₂ MW: 174.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00236227 CAS Number: 125903-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...