4-Methoxy-1-butanol 98+%

Supplier: Thermo Scientific Chemicals
Warning

H50339-03 H50339-06
CAAAAH50339-03EA 73.94 CAD
CAAAAH50339-03 CAAAAH50339-06
4-Methoxy-1-butanol 98+%
4-Methoxy-1-butanol

Purity: 98+% CAS No.: 111-32-0 Molecular Formula: C5H12O2 Formula Weight: 104.15 Flash Point: 73°(163°F) Odor: Mild Boiling Point: 66°/7mm MDL No.: MFCD00040437

Formula: C₅H₁₂O₂
Boiling Pt: 66 °C (7 mmHg)
Density: 0,93
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040437
CAS Number: 111-32-0
EINECS: 203-858-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR