4-(1-Aminoethyl)pyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Pyridin-4-ylethylamine, 1-(4-Pyridyl)ethylamine

H50303-03 H50303-MD
CAAAAH50303-03EA 167.48 CAD
CAAAAH50303-03 CAAAAH50303-MD
4-(1-Aminoethyl)pyridine 97%
4-(1-Aminoethyl)pyridine

Purity: 97% CAS No.: 50392-78-4 Molecular Formula: C7H10N2 Formula Weight: 122.17 UN No.: 2735 MDL No.: MFCD02245004

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02245004
CAS Number: 50392-78-4
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR