Ethyl-6-aminohexanoate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
6-Aminohexanoic acid ethyl ester, 6-Aminocaproic acid ethyl ester
H50296-03
H50296-MD
CAAAAH50296-03EA
393.42
CAD
CAAAAH50296-03
CAAAAH50296-MD
Ethyl-6-aminohexanoate 98%
Ethyl 6-aminohexanoate
Purity: 98%
CAS No.: 371-34-6
Molecular Formula: C8H17NO2
Formula Weight: 159.23
MDL No.: MFCD00233495
Formula:
C₈H₁₇NO₂ MW: 159.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00233495 CAS Number: 371-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...