1-(3-Hydroxypropyl)piperazine 98%

Supplier: Thermo Scientific Chemicals
Danger

H50290-03 H50290-06
CAAAAH50290-03EA 126.4 CAD
CAAAAH50290-03 CAAAAH50290-06
1-(3-Hydroxypropyl)piperazine 98%
1-(3-Hydroxypropyl)piperazine

Purity: 98% CAS No.: 5317-32-8 Molecular Formula: C7H16N2O Formula Weight: 144.22 Melting Point: 48-52° MDL No.: MFCD00023132

Formula: C₇H₁₆N₂O
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00023132
CAS Number: 5317-32-8
EINECS: 226-176-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR