3-(4-Chlorophenyl)propionic acid 94%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4-Chlorophenyl)propanoic acid

H50204-03 H50204-06
CAAAAH50204-03EA 71.57 CAD
CAAAAH50204-03 CAAAAH50204-06
3-(4-Chlorophenyl)propionic acid 94%
3-(4-Chlorophenyl)propionic acid

Purity: 94% CAS No.: 2019-34-3 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Melting Point: 118-120° MDL No.: MFCD00016555

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00016555
CAS Number: 2019-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR