2-Chlorothiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

H50155-03 H50155-06
CAAAAH50155-03EA 34.6 CAD
CAAAAH50155-03 CAAAAH50155-06
2-Chlorothiazole 97%
2-Chlorothiazole

Purity: 97% CAS No.: 3034-52-4 Molecular Formula: C3H2ClNS Formula Weight: 119.57 MDL No.: MFCD00210701

Formula: C₃H₂ClNS
Storage Temperature: Ambient
MDL Number: MFCD00210701
CAS Number: 3034-52-4
EINECS: 221-228-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR