2-Chloro-4-iodo-3-methylpyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-4-iodo-3-picoline

H50107-03 H50107-06
CAAAAH50107-03EA 208.56 CAD
CAAAAH50107-03 CAAAAH50107-06
2-Chloro-4-iodo-3-methylpyridine 95%
2-Chloro-4-iodo-3-methylpyridine

Purity: 95% CAS No.: 153034-88-9 Molecular Formula: C6H5ClIN Formula Weight: 253.47 Melting Point: 106-107° MDL No.: MFCD03095293

Formula: C₆H₅ClIN
MW: 253.47
Melting Pt: 106…107 °C
Storage Temperature: Ambient
MDL Number: MFCD03095293
CAS Number: 153034-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR