4-Amino-3-iodopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H50027-03 H50027-06
CAAAAH50027-03EA 195.92 CAD
CAAAAH50027-03 CAAAAH50027-06
4-Amino-3-iodopyridine 97%
4-Amino-3-iodopyridine

Purity: 97% CAS No.: 88511-27-7 Molecular Formula: C5H5IN2 Formula Weight: 220.01 MDL No.: MFCD02663853

Formula: C₅H₅IN₂
Storage Temperature: Ambient
MDL Number: MFCD02663853
CAS Number: 88511-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR