trans-4-Methylcyclohexanecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: trans-4-Methyl-1-cyclohexanecarboxylic acid

H27455-03 H27455-06
CAAAH27455-03EA 195.92 CAD
CAAAH27455-03 CAAAH27455-06
trans-4-Methylcyclohexanecarboxylic acid 98%
trans-4-Methylcyclohexanecarboxylic acid

Purity: 98% CAS No.: 13064-83-0 Molecular Formula: C8H14O2 Formula Weight: 142.20 Melting Point: 109-110° MDL No.: MFCD00074943

Formula: C₈H₁₄O₂
Melting Pt: 109…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00074943
CAS Number: 13064-83-0
EINECS: 235-959-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR