2-Methyl-1-butene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H27230-06
H27230-14
CAAAH27230-06EA
97.96
CAD
CAAAH27230-06
CAAAH27230-14
2-Methyl-1-butene 98%
2-Methyl-1-butene
Purity: 98%
CAS No.: 563-46-2
Molecular Formula: C5H10
Formula Weight: 70.13
Storage Temperature: 2°C to 8°C
Flash Point: -48°(-54°F)
Refractive Index: 1.3740
Melting Point: -137°
Boiling Point: 31-32°
UN No.: 2459
MDL No.: MFCD00009333
Formula:
C₅H₁₀ MW: 70.13 g/mol Boiling Pt: 31…32 °C Melting Pt: –137 °C Density: 0.651 Flash Pt: –48 °C (–54 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009333 CAS Number: 563-46-2 EINECS: 209-250-7 UN: 2459 ADR: 3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...