D-allo-Threonine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-D-Allo-Thr-OH, (2R,3R)-2-Amino-3-hydroxybutanoic acid
H27050-03
H27050-MD
CAAH27050-03EA
483.48
CAD
CAAH27050-03
CAAH27050-MD
D-allo-Threonine 99%
D-allo-Threonine
Purity: 99%
CAS No.: 24830-94-2
Molecular Formula: C4H9NO3
Formula Weight: 119.12
Melting Point: 268° dec.
Beilstein Reference: 1721644
Formula:
C₄H₉NO₃ Melting Pt: 268 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004526 CAS Number: 24830-94-2 EINECS: 246-488-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...