2-Bromo-6-(trifluoromethyl)pyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H25749-03
H25749-06
CAAAH25749-03EA
205.4
CAD
CAAAH25749-03
CAAAH25749-06
CAAAH25749-MD
2-Bromo-6-(trifluoromethyl)pyridine 97%
2-Bromo-6-(trifluoromethyl)pyridine
Purity: 97%
CAS No.: 189278-27-1
Molecular Formula: C6H3BrF3N
Formula Weight: 226.00
Melting Point: 48-51°
Boiling Point: 78-79°/14mm
MDL No.: MFCD00153087
Formula:
C₆H₃BrF₃N Boiling Pt: 78…79 °C/14 mm Melting Pt: 48…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153087 CAS Number: 189278-27-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...