2-Bromo-4-(trifluoromethyl)pyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H25748-03
H25748-06
CAAAH25748-03EA
278.08
CAD
CAAAH25748-03
CAAAH25748-06
CAAAH25748-MD
2-Bromo-4-(trifluoromethyl)pyridine 97%
2-Bromo-4-(trifluoromethyl)pyridine
Purity: 97%
CAS No.: 175205-81-9
Molecular Formula: C6H3BrF3N
Formula Weight: 226.00
Flash Point: 73°(164°F)
Refractive Index: 1.4780
Boiling Point: 84-85°/14mm
MDL No.: MFCD00153085
Formula:
C₆H₃BrF₃N Boiling Pt: 84…85 °C/14 mm Density: 1.83 Flash Pt: 73 °C(164 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00153085 CAS Number: 175205-81-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...