2-Bromo-5-nitrobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25704-06
B25704-14
CAAAAB25704-03EA
49.45
CAD
CAAAAB25704-03
CAAAAB25704-06
CAAAAB25704-14
2-Bromo-5-nitrobenzoic acid 98%
2-Bromo-5-nitrobenzoic acid
Purity: 98%
CAS No.: 943-14-6
Molecular Formula: C7H4BrNO4
Formula Weight: 246.02
Melting Point: 178-181°
Beilstein Reference: 980242
Formula:
C₇H₄BrNO₄ Boiling Pt: 371 °C (1013 hPa) Melting Pt: 178…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134558 CAS Number: 943-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...