5-Bromo-o-anisidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2-anisidine, 5-Bromo-2-methoxyaniline, 2-Amino-4-bromoanisole

B25693-06 B25693-14
CAAAAB25693-06EA 164.32 CAD
CAAAAB25693-06 CAAAAB25693-14
5-Bromo-o-anisidine 97%
5-Bromo-o-anisidine

Purity: 97% CAS No.: 6358-77-6 Molecular Formula: C7H8BrNO Formula Weight: 202.05 Melting Point: 94-96° UN No.: 2811 MDL No.: MFCD04037882

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD04037882
CAS Number: 6358-77-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR