2',6'-Dimethylacetanilide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2,6-Dimethylphenyl)acetamide, 2,6-Dimethylacetanilide

B25687-14 B25687-22
CAAAAB25687-14EA 58.93 CAD
CAAAAB25687-14 CAAAAB25687-22
2',6'-Dimethylacetanilide 97%
2',6'-Dimethylacetanilide

Purity: 97% CAS No.: 2198-53-0 Molecular Formula: C10H13NO Formula Weight: 163.22 Melting Point: 179-183° MDL No.: MFCD00008675

Formula: C₁₀H₁₃NO
MW: 163.22 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00008675
CAS Number: 2198-53-0
EINECS: 218-596-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR