3-Methyl-2-nitrobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Nitro-m-toluic acid, 3-Methyl-2-nitrobenzoic acid

B25682-14 B25682-22
CAAAAB25682-14EA 41.71 CAD
CAAAAB25682-14 CAAAAB25682-22
3-Methyl-2-nitrobenzoic acid 98%
2-Nitro-m-toluic acid

Purity: 98% CAS No.: 5437-38-7 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 219-223° MDL No.: MFCD00007180

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 340
Melting Pt: 219…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00007180
CAS Number: 5437-38-7
EINECS: 226-610-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR