(3,5-Dimethoxyphenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B25672-03 B25672-06
CAAAAB25672-03EA 108.23 CAD
CAAAAB25672-03 CAAAAB25672-06
(3,5-Dimethoxyphenyl)acetic acid 98%
(3,5-Dimethoxyphenyl)acetic acid

Purity: 98% CAS No.: 4670-10-4 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 102-104° MDL No.: MFCD00016827

Formula: (C₆H₃(OCH₃)₂)CH₂COOH
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00016827
CAS Number: 4670-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR