5-Phenyl-1H-tetrazole 99%

Supplier: Thermo Scientific Chemicals
Warning

B25664-14 B25664-22
CAAAAB25664-14EA 84.85 CAD
CAAAAB25664-14 CAAAAB25664-22
5-Phenyl-1H-tetrazole 99%
5-Phenyl-1H-tetrazole

Purity: 99% CAS No.: 18039-42-4 Molecular Formula: C7H6N4 Formula Weight: 146.15 Odor: Odorless Melting Point: ca 215° dec. MDL No.: MFCD00022388

Formula: C₇H₆N₄
MW: 146.15 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: ∼ 215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00022388
CAS Number: 18039-42-4
EINECS: 241-950-8
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR