(Trifluoromethoxy)benzene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B25658-14
B25658-22
CAAAAB25658-14EA
84.06
CAD
CAAAAB25658-14
CAAAAB25658-22
(Trifluoromethoxy)benzene 99%
(Trifluoromethoxy)benzene
Purity: 99%
CAS No.: 456-55-3
Molecular Formula: C7H5F3O
Formula Weight: 162.11
Flash Point: 12°(53°F)
Refractive Index: 1.4060
Odor: Aromatic
Boiling Point: 102-104°
Beilstein Reference: 2043132
UN No.: 1993
MDL No.: MFCD00040832
Formula:
C₇H₅F₃O Boiling Pt: 102…104 °C Melting Pt: –50 °C Density: 1.226 Flash Pt: 12 °C (53 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040832 CAS Number: 456-55-3 EINECS: 207-269-5 UN: 1993 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...