9-Phenanthrenecarbaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

B25648-03 B25648-06
CAAAAB25648-03EA 88.95 CAD
CAAAAB25648-03 CAAAAB25648-06
9-Phenanthrenecarbaldehyde 97%
9-Phenanthrenecarbaldehyde

Purity: 97% CAS No.: 4707-71-5 Molecular Formula: C15H10O Formula Weight: 206.24 Melting Point: 100-103° MDL No.: MFCD00001175

Formula: C₁₅H₁₀O
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00001175
CAS Number: 4707-71-5
EINECS: 225-194-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR