5-Fluoro-2-methylanisole 98%
Supplier: Thermo Scientific Chemicals
B25629-03
B25629-06
CAAAAB25629-03EA
42.03
CAD
CAAAAB25629-03
CAAAAB25629-06
5-Fluoro-2-methylanisole 98%
5-Fluoro-2-methylanisole
Purity: 98%
CAS No.: 95729-22-9
Molecular Formula: C8H9FO
Formula Weight: 140.16
Refractive Index: 1.4940
Formula:
C₈H₉FO Storage Temperature: Ambient |
MDL Number:
MFCD00673008 CAS Number: 95729-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...