3-Acetylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3'-Acetophenonecarboxylic acid, 3-Acetylbenzoic Acid

B25616-03 B25616-06
CAAAAB25616-03EA 162.74 CAD
CAAAAB25616-03 CAAAAB25616-06
3-Acetylbenzoic acid 98%
3-Acetylbenzoic acid

Purity: 98% CAS No.: 586-42-5 Molecular Formula: C9H8O3 Formula Weight: 164.16 Odor: Odorless Melting Point: 162-166° MDL No.: MFCD00045847

Formula: C₉H₈O₃
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 162…166 °C
Density: 1,23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00045847
CAS Number: 586-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR