Bis(4-chlorophenyl) disulfide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bis(4-chlorophenyl) disulphide, Bis(4-chlorophenyl) disulfide
B25604-06
B25604-14
CAAAAB25604-06EA
94.48
CAD
CAAAAB25604-06
CAAAAB25604-14
Bis(4-chlorophenyl) disulfide 98+%
Bis(4-chlorophenyl) disulphide
Purity: 98+%
CAS No.: 1142-19-4
Molecular Formula: C12H8Cl2S2
Formula Weight: 287.23
Melting Point: 71-74°
Beilstein Reference: 1644663
Formula:
C₁₂H₈Cl₂S₂ MW: 287.23 g/mol Melting Pt: 71…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013642 CAS Number: 1142-19-4 EINECS: 214-531-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...