2,4-Difluoro-3-methoxybenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25597-06
B25597-14
CAAAAB25597-06EA
118.97
CAD
CAAAAB25597-06
CAAAAB25597-14
2,4-Difluoro-3-methoxybenzoic acid 98+%
2,4-Difluoro-3-methoxybenzoic acid
Purity: 98+%
CAS No.: 178974-97-5
Molecular Formula: C8H6F2O3
Formula Weight: 188.13
Melting Point: 193-195°
MDL No.: MFCD02258905
Formula:
C₈H₆F₂O₃ Melting Pt: 193…195 °C Storage Temperature: Ambient |
MDL Number:
MFCD02258905 CAS Number: 178974-97-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...