3',5'-Dihydroxyacetophenone 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(3,5-Dihydroxyphenyl)ethan-1-one, 1-(3,5-Dihydroxyphenyl)ethanone

B25567-09 B25567-18
CAAAAB25567-09EA 124.5 CAD
CAAAAB25567-09 CAAAAB25567-18
3',5'-Dihydroxyacetophenone 96%
3',5'-Dihydroxyacetophenone

Purity: 96% CAS No.: 51863-60-6 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 142-149° Beilstein Reference: 1936502

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 142…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00002290
CAS Number: 51863-60-6
EINECS: 257-480-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR