5,6-Dimethoxy-1-indanone 97%

Supplier: Thermo Scientific Chemicals
Warning

B25559-06 B25559-14
CAAAAB25559-06EA 184.86 CAD
CAAAAB25559-06 CAAAAB25559-14
5,6-Dimethoxy-1-indanone 97%
5,6-Dimethoxy-1-indanone

Purity: 97% CAS No.: 2107-69-9 Molecular Formula: C11H12O3 Formula Weight: 192.21 Melting Point: 118-120° MDL No.: MFCD00003790

Formula: C₁₁H₁₂O₃
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00003790
CAS Number: 2107-69-9
EINECS: 218-287-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR