1-Fluoro-3-nitrobenzene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Fluoronitrobenzene

B25553-14 B25553-22
CAAAAB25553-14EA 104.44 CAD
CAAAAB25553-14 CAAAAB25553-22
1-Fluoro-3-nitrobenzene 98+%
1-Fluoro-3-nitrobenzene

Purity: 98+% CAS No.: 402-67-5 Molecular Formula: C6H4FNO2 Formula Weight: 141.10 Flash Point: 76°(168°F) Refractive Index: 1.5250 Melting Point: 2° Boiling Point: 204-206° Beilstein Reference: 1862210 UN No.: 2810 MDL No.: MFCD00007196

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 204…206 °C
Melting Pt: 2 °C
Density: 1.325
Flash Pt: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007196
CAS Number: 402-67-5
EINECS: 206-953-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR