Methyl-4-iodo-3-nitrobenzoate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25544-06
B25544-14
CAAAAB25544-06EA
148.84
CAD
CAAAAB25544-06
CAAAAB25544-14
Methyl-4-iodo-3-nitrobenzoate 99%
Methyl 4-iodo-3-nitrobenzoate
Purity: 99%
CAS No.: 89976-27-2
Molecular Formula: C8H6INO4
Formula Weight: 307.05
Melting Point: 101-104°
MDL No.: MFCD08064223
Formula:
C₈H₆INO₄ Melting Pt: 101…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD08064223 CAS Number: 89976-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...