4-Bromo-m-anisidine 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromo-3-methoxyaniline, 5-Amino-2-bromoanisole, 4-Bromo-3-anisidine
B25541-06
B25541-14
CAAAAB25541-06EA
178.54
CAD
CAAAAB25541-06
CAAAAB25541-14
4-Bromo-m-anisidine 97+%
4-Bromo-m-anisidine
Purity: 97+%
CAS No.: 19056-40-7
Molecular Formula: C7H8BrNO
Formula Weight: 202.05
Melting Point: 96-100°
Beilstein Reference: 2690835
UN No.: 2811
MDL No.: MFCD05664063
Formula:
C₇H₈BrNO MW: 202.05 g/mol Melting Pt: 96…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664063 CAS Number: 19056-40-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...